Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

393444

Sigma-Aldrich

Copper(II) sulfate on alumina

extent of labeling: 9 wt. % loading, matrix neutral alumina support

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4
CAS Number:
Molecular Weight:
161.62
MDL number:
UNSPSC Code:
12352300

extent of labeling

9 wt. % loading

matrix

neutral alumina support

SMILES string

[Cu++].[O-]S([O-])(=O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric J Wagner et al.
Journal of aquatic animal health, 25(2), 125-130 (2013-05-24)
Tests were conducted to determine the concentrations of copper sulfate needed to kill Flavobacterium psychrophilum, the cause of bacterial coldwater disease, either in vitro or on Rainbow Trout Oncorhynchus mykiss eggs. For the in vitro test, a plastic strip dipped
Eun A Choi et al.
International journal of molecular medicine, 31(1), 154-162 (2012-11-07)
Egg white-chalcanthite (EWCC) is a mixture of egg white and chalcanthite prepared by roasting chalcanthite (which is a natural mineral mainly composed of CuSO4•5H2O) to the point of dehydration, pulverizing the dehydrated chalcanthite and then mixing the pulverized chalcanthite to
Xu-Chen Cao et al.
PloS one, 8(2), e56735-e56735 (2013-03-08)
The aquaporin (AQP) family consists of a number of small integral membrane proteins that transport water and glycerol. AQPs are critical for trans-epithelial fluid transport. Recent reports demonstrated that AQPs, particularly AQP1 and AQP5, are expressed in high grade tumor
M Seder-Colomina et al.
Ecotoxicology (London, England), 22(1), 199-205 (2012-11-10)
Microbial mats are coastal ecosystems that consist mainly of cyanobacteria, primary producers in these habitats that play an important role in stabilising delta sediments. However, these ecosystems are subject to various kinds of pollution, including metal contamination, placing their survival
Ashlie Watters et al.
Biofouling, 29(1), 21-28 (2012-12-01)
Quagga mussels (Dreissena rostriformis bugensis) have created economic and ecological impacts in the western United States since their discovery in 2007. This study focuses on chemical control for preventing the spread of these mussels. The effectiveness of EarthTec(®) in killing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service