Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

388335

Sigma-Aldrich

Palmityl palmitate

98%

Synonym(s):

Cetyl palmitate, Hexadecyl hexadecanoate, Palmitic acid palmityl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2(CH2)15CH3
CAS Number:
Molecular Weight:
480.85
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

98%

mp

55-56 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

InChI

1S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3

Inchi Key

PXDJXZJSCPSGGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Blasi et al.
International journal of pharmaceutics, 419(1-2), 287-295 (2011-08-11)
The aim of this study was to optimize the formulation of lipid nanoparticles (NPs), intended for brain targeting, with the aid of a computer generated experimental design. The high pressure homogenization technique, selected for this purpose, was suitable to formulate
G Lukowski et al.
International journal of pharmaceutics, 196(2), 201-205 (2000-03-04)
Solid lipid nanoparticles (SLN) as alternative intravenous colloidal drug carriers were produced by high pressure homogenisation of the melted lipid cetylpalmitate. The crystallographic properties of the used cetylpalmitate SLN were characterised by small angle X-ray scattering (SAXS) and X-ray diffraction
Veerawat Teeranachaideekul et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 67(1), 141-148 (2007-03-10)
In the present study, nanostructured lipid carriers (NLC) composed of cetyl palmitate with various amounts of caprylic/capric triacylglycerols (as liquid lipid) were prepared and Coenzyme Q(10) (Q(10)) has been incorporated in such carriers due to its high lipophilic character. A
N Schöler et al.
International journal of pharmaceutics, 196(2), 235-239 (2000-03-04)
In order to investigate the interaction of preserved solid lipid nanoparticles (SLN) with murine peritoneal macrophages (Mpsi), cytotoxicity and proinflammatory effects of two different solid lipid nanoparticles (SLN) preparations consisting of either compritol (CO) or cetyl palmitate (CP) preserved with
Effat Sadat Farboud et al.
International journal of nanomedicine, 6, 611-617 (2011-06-16)
Solid lipid nanoparticles (SLNs) of coenzyme Q10 (CoQ10) were formulated by a high-pressure homogenization method. The best formulation of SLN dispersion consisted of 13% lipid (cetyl palmitate or stearic acid), 8% surfactant (Tween 80 or Tego Care 450), and water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service