Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

385328

Sigma-Aldrich

trans-1-(2-Naphthyl)-3-phenyl-2-propen-1-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14O
CAS Number:
Molecular Weight:
258.31
MDL number:
UNSPSC Code:
12352103

assay

98%

SMILES string

O=C(\C=C\c1ccccc1)c2ccc3ccccc3c2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service