About This Item
assay
99%
SMILES string
C1CCCOCCC1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
208.4 °F - closed cup
flash_point_c
98.00 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service