381152
Ruthenium on alumina
extent of labeling: 5 wt. % loading, powder, Degussa type H213 R/D
About This Item
Recommended Products
form
powder
reaction suitability
reagent type: catalyst
core: ruthenium
extent of labeling
5 wt. % loading
SMILES string
[Ru]
InChI
1S/Ru
InChI key
KJTLSVCANCCWHF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service