Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

379638

Sigma-Aldrich

Dinitrato(1,10-phenanthroline)copper(II)

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10CuN4O6
CAS Number:
Molecular Weight:
369.78
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

96%

SMILES string

[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.c1cnc2c(c1)ccc3cccnc23


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service