377678
[(S)-(−)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene]chloro(p-cymene)ruthenium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C54H46Cl2P2Ru
CAS Number:
Molecular Weight:
928.87
MDL number:
UNSPSC Code:
12352000
SMILES string
[Cl-].Cl[Ru+].CC(C)c1ccc(C)cc1.c2ccc(cc2)P(c3ccccc3)c4ccc5ccccc5c4-c6c(ccc7ccccc67)P(c8ccccc8)c9ccccc9
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service