Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

375497

Sigma-Aldrich

Phenoxyacetaldehyde dimethyl acetal

99%

Synonym(s):

(2,2-Dimethoxyethoxy)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH(OCH3)2
CAS Number:
Molecular Weight:
182.22
Beilstein/REAXYS Number:
5809638
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

78 °C/0.6 mmHg (lit.)

density

1.076 g/mL at 25 °C (lit.)

functional group

acetal
ether
phenoxy

SMILES string

COC(COc1ccccc1)OC

InChI

1S/C10H14O3/c1-11-10(12-2)8-13-9-6-4-3-5-7-9/h3-7,10H,8H2,1-2H3

InChI key

QHMTZLHLQBIYJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenoxyacetaldehyde dimethyl acetal may be used in chemical synthesis.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service