All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H20O2Si
CAS Number:
Molecular Weight:
188.34
Beilstein/REAXYS Number:
1930256
EC Number:
MDL number:
UNSPSC Code:
12352100
assay
97%
refractive index
n20/D 1.418 (lit.)
bp
67 °C/13 mmHg (lit.)
density
0.86 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)OC(=O)C[Si](C)(C)C
InChI
1S/C9H20O2Si/c1-9(2,3)11-8(10)7-12(4,5)6/h7H2,1-6H3
InChI key
HOHBQWITMXOSOW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service