Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

374237

Sigma-Aldrich

CHAPSO

95%

Synonym(s):

3-([3-Cholamidopropyl]dimethylammonio)-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
Beilstein/REAXYS Number:
5842642
MDL number:
UNSPSC Code:
12352103

assay

95%

optical activity

[α]20/D +19°, c = 2 in H2O

mol wt

micellar avg mol wt 7000

aggregation number

11

CMC

8 mM (20-25°C)

mp

184-186 °C (lit.)

transition temp

cloud point 90 °C

SMILES string

[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@]([H])(CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(=O)NCCC[N+](C)(C)CC(O)CS([O-])(=O)=O

InChI

1S/C32H58N2O8S/c1-20(7-10-29(39)33-13-6-14-34(4,5)18-23(36)19-43(40,41)42)24-8-9-25-30-26(17-28(38)32(24,25)3)31(2)12-11-22(35)15-21(31)16-27(30)37/h20-28,30,35-38H,6-19H2,1-5H3,(H-,33,39,40,41,42)/t20-,21+,22-,23?,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

GUQQBLRVXOUDTN-XOHPMCGNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A nondenaturing zwitterionic detergent with characteristics similar to CHAPS, although it is more soluble due to a more polar head group. Useful for the solubilization of integral membrane proteins.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service