Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

373877

Sigma-Aldrich

Glycerol propoxylate-block-ethoxylate

average Mn ~5,200

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H22O7
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

>1 (vs air)

vapor pressure

<0.3 mmHg ( 20 °C)

description

functionality 2.5

mol wt

average Mn ~5,200

contains

400-800 ppm BHT as inhibitor

composition

ethoxylate, 5%
propoxylate, 95%

density

1.02 g/mL at 25 °C

SMILES string

OCCO.CC(O)CO.OCC(O)CO

InChI

1S/C3H8O3.C3H8O2.C2H6O2/c4-1-3(6)2-5;1-3(5)2-4;3-1-2-4/h3-6H,1-2H2;3-5H,2H2,1H3;3-4H,1-2H2

InChI key

VKVYTKULMKDWLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service