373559
2-Chlorophenothiazine
95%
Synonym(s):
2-Chlorothiodiphenylamine
About This Item
Recommended Products
assay
95%
mp
195-200 °C (lit.)
SMILES string
Clc1ccc2Sc3ccccc3Nc2c1
InChI
1S/C12H8ClNS/c13-8-5-6-12-10(7-8)14-9-3-1-2-4-11(9)15-12/h1-7,14H
Inchi Key
KFZGLJSYQXZIGP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
replaced by
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service