Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

371955

Sigma-Aldrich

2,2,3,4,4,4-Hexafluoro-1-butanol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(F)CF2CH2OH
CAS Number:
Molecular Weight:
182.06
Beilstein/REAXYS Number:
1753872
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95%

refractive index

n20/D 1.312 (lit.)

bp

114 °C/740 mmHg (lit.)

density

1.557 g/mL at 25 °C (lit.)

SMILES string

OCC(F)(F)C(F)C(F)(F)F

InChI

1S/C4H4F6O/c5-2(4(8,9)10)3(6,7)1-11/h2,11H,1H2

InChI key

LVFXLZRISXUAIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,3,4,4,4-Hexafluoro-1-butanol may be used in the preparation of fluorinated cationic guar gum (FCGG).[1],[2] It may be employed as solvent to evaluate the dynamic nuclear polarization (DNP) parameters for the solutions of free radicals Galvinoxyl (GALV) and α,γ-bisdiphenylene-β-phenyl allyl complex with benzene (1:1) (BDPA).[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Wang et al.
Carbohydrate polymers, 95(2), 637-643 (2013-05-08)
The fluorinated cationic guar gum (FCGG) was prepared with cationic guar gum (CGG) and fluorinated reactive monomer (FAM). FAM was synthesized from isophorone diisocyanate and 2,2,3,4,4,4-hexafluoro-1-butanol. The molecular structure, associating and rheological behaviors of FCGG were studied by the FT-IR
A low field fluorine-electron double resonance study for GALV and BDPA in some aliphatic and aromatic solvents.
Peksoz A, et al.
Zeitschrift fur Naturforschung, 64, 477-484 (2009)
Chen Wang et al.
Carbohydrate polymers, 99, 638-645 (2013-11-28)
The interactions between the fluorinated cationic guar gum (FCGG) and ionic surfactants including cetyl trimethyl ammonium bromide (CTAB) and sodium lauryl sulfate (SDS) were studied by light scattering, fluorescence spectroscopy, UV-spectrophotometer, (19)F NMR and dynamic rheometer, respectively. The FCGG is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service