Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369276

Sigma-Aldrich

Lithium trimethylsilanolate solution

1.0 M in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOLi
CAS Number:
Molecular Weight:
96.13
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

vapor pressure

24.12 psi ( 55 °C)
6.7 psi ( 20 °C)

concentration

1.0 M in methylene chloride

bp

40 °C

density

1.278 g/mL at 25 °C

SMILES string

[Li+].C[Si](C)(C)[O-]

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grzegorz Piechota et al.
Bioresource technology, 103(1), 16-20 (2011-10-29)
The removal and determination of trimethylsilanol (TMSOH) in landfill gas has been studied before and after the special E3000-ITC System. The system works according to principle of temperature swing. The performance of TMSOH and humidity removal was 20% and more
A Isquith et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(3), 263-266 (1988-03-01)
Six organosilicon compounds which had been found to have clastogenic activity in an in vitro battery of genotoxicity assays were evaluated in rat bone marrow cytogenetic assays for assessing clastogenicity in an in vivo system. None of the six compounds
D M Hui et al.
Clinica chimica acta; international journal of clinical chemistry, 302(1-2), 171-188 (2000-11-14)
We developed a new assay method for fluoride anion (F(-)) a specific metabolite of sarin. Trimethyifluorosilane (TMFS) was derivatized from F(-) with trimethylsilanol, and TMFS was detected with a GC-flame ionization detector (FID) and capillary column system. The linear range
Todd A Duncombe et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13765-13770 (2012-09-01)
We introduce the wetting barrier ratchet, a digital microfluidic technology for directed drop transport in an open air environment. Cyclic drop footprint oscillations initiated by orthogonal vibrations as low as 37 μm in amplitude at 82 Hz are rectified into
Fabricio A Hansel et al.
Rapid communications in mass spectrometry : RCM, 25(13), 1893-1898 (2011-06-04)
A methodology is presented for the determination of dihydroxy fatty acids preserved in the 'bound' phase of organic residues preserved in archaeological potsherds. The method comprises saponification, esterification, silica gel column chromatographic fractionation, and analysis by gas chromatography/mass spectrometry. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service