368970
(2-Bromoethyl) chloroformate
technical grade, 90%
About This Item
grade
technical grade
assay
90%
refractive index
n20/D 1.478 (lit.)
density
1.749 g/mL at 25 °C (lit.)
application(s)
peptide synthesis
SMILES string
ClC(=O)OCCBr
InChI
1S/C3H4BrClO2/c4-1-2-7-3(5)6/h1-2H2
InChI key
ZLCKDYMZBZNBMJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
143.6 °F - closed cup
flash_point_c
62 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service