Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

367192

Sigma-Aldrich

Nickel(II) peroxide hydrate

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NiO2 · xH2O
CAS Number:
Molecular Weight:
90.69 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

form

powder

mol wt

Mw 90.69 g/mol (anyhydrous basis)

reaction suitability

reagent type: catalyst
core: nickel

reagent type: oxidant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

application(s)

battery manufacturing

greener alternative category

SMILES string

O.O1O[Ni]1

InChI

1S/Ni.O2.H2O/c;1-2;/h;;1H2/q+2;-2;

InChI key

WFGXJOHFKNQUMN-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

signalword

Danger

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Ox. Sol. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service