Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

36651

Sigma-Aldrich

N,N′-Dicyclohexylcarbodiimide solution

purum, ~1 M in NMP

Synonym(s):

DCC solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N=C=NC6H11
CAS Number:
Molecular Weight:
206.33
Beilstein/REAXYS Number:
610662
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

concentration

~1 M in NMP

refractive index

n20/D 1.480

SMILES string

C1CCC(CC1)N=C=NC2CCCCC2

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

203.0 °F

flash_point_c

95 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siobhan P Brown et al.
American heart journal, 169(3), 334-341 (2015-03-03)
The Resuscitation Outcomes Consortium is conducting a randomized trial comparing survival with hospital discharge after continuous chest compressions without interruption for ventilation versus currently recommended American Heart Association cardiopulmonary resuscitation with interrupted chest compressions in adult patients with out-of-hospital cardiac
Denys Pogoryelov et al.
Nature chemical biology, 6(12), 891-899 (2010-10-26)
The microscopic mechanism of coupled c-ring rotation and ion translocation in F(1)F(o)-ATP synthases is unknown. Here we present conclusive evidence supporting the notion that the ability of c-rings to rotate within the F(o) complex derives from the interplay between the
Masashi Toei et al.
The Journal of biological chemistry, 288(36), 25717-25726 (2013-07-31)
N,N-Dicyclohexylcarbodiimide (DCCD) is a classical inhibitor of the F0F1-ATP synthase (F0F1), which covalently binds to the highly conserved carboxylic acid of the proteolipid subunit (c subunit) in F0. Although it is well known that DCCD modification of the c subunit
Kenji Mizutani et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(33), 13474-13479 (2011-08-05)
The prokaryotic V-ATPase of Enterococcus hirae, closely related to the eukaryotic enzymes, provides a unique opportunity to study the ion-translocation mechanism because it transports Na(+), which can be detected by radioisotope (22Na(+)) experiments and X-ray crystallography. In this study, we
Marten Staal et al.
Plant physiology, 155(4), 2049-2055 (2011-02-02)
In Arabidopsis (Arabidopsis thaliana; Columbia-0) roots, the so-called zone of cell elongation comprises two clearly different domains: the transition zone, a postmeristematic region (approximately 200-450 μm proximal of the root tip) with a low rate of elongation, and a fast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service