Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

366293

Sigma-Aldrich

1-Isothiocyanato-4-(trans-4-propylcyclohexyl)benzene

liquid crystal (nematic), 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C6H10C6H4NCS
CAS Number:
Molecular Weight:
259.41
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

99%

form

liquid crystal (nematic)

mp

41-44 °C (lit.)

transition temp

crystalline phase to nematic phase 38.5 °C
nematic phase to isotropic phase 41.5 °C

SMILES string

CCC[C@H]1CC[C@@H](CC1)c2ccc(cc2)N=C=S

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service