assay
99%
mp
182-183 °C (lit.)
functional group
ester
SMILES string
COC(=O)c1cc2c(OC)cc(OC)cc2[nH]1
InChI
1S/C12H13NO4/c1-15-7-4-9-8(11(5-7)16-2)6-10(13-9)12(14)17-3/h4-6,13H,1-3H3
InChI key
TVUJONIYWNBWPM-UHFFFAOYSA-N
General description
Methyl 4,6-dimethoxy-2-indolecarboxylate is an indole derivative.
Application
Methyl 4,6-dimethoxy-2-indolecarboxylate may be used as reactant:
- in the nitration reactions with nitric acid
- in the preparation of light-dependent tumor necrosis factor-α inhibitors
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bioorganic & Medicinal Chemistry, 13, 533-533 (2003)
Tetrahedron, 60, 10779-10779 (2004)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service