About This Item
Recommended Products
assay
96%
impurities
~4% dimethyl disulfide
refractive index
n20/D 1.49 (lit.)
bp
110 °C (lit.)
density
1.288 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CSC(Cl)=O
InChI
1S/C2H3ClOS/c1-5-2(3)4/h1H3
InChI key
YPSUCTSXOROPBS-UHFFFAOYSA-N
accessory
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 2 Inhalation
target_organs
Upper respiratory tract
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
87.8 °F - closed cup
flash_point_c
31 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service