Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

364908

Sigma-Aldrich

(S)-(−)-1,1-Diphenyl-1,2-propanediol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)C(C6H5)2OH
CAS Number:
Molecular Weight:
228.29
Beilstein/REAXYS Number:
3202232
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

99%

optical activity

[α]25/D −100°, c = 1 in methanol

mp

89-92 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

C[C@H](O)C(O)(c1ccccc1)c2ccccc2

InChI

1S/C15H16O2/c1-12(16)15(17,13-8-4-2-5-9-13)14-10-6-3-7-11-14/h2-12,16-17H,1H3/t12-/m0/s1

InChI key

RQKXFLUAQLDHMO-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chiral glycol for the synthesis of chiral sulfoxides and epoxides.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 23, 45-45 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service