All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10O2S2
CAS Number:
Molecular Weight:
178.27
MDL number:
UNSPSC Code:
12352103
assay
95%
refractive index
n20/D 1.548 (lit.)
density
1.17 g/mL at 25 °C (lit.)
SMILES string
CCOC(=S)SCC(C)=O
Application
Highly enolizable precursor to some ketene hemithioacetals, 2-aminothiazoles, and free-radical probes.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the Chemical Society. Perkin Transactions 1, 1623-1623 (1985)
Journal of the Chemical Society. Perkin Transactions 1, 1603-1603 (1975)
Journal of the Chemical Society. Perkin Transactions 1, 39-39 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service