Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

362611

Sigma-Aldrich

Dichloro[(R)-N,N-dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine]palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H21Cl2FeNPPd
CAS Number:
Molecular Weight:
611.60
MDL number:
UNSPSC Code:
12352103

specific activity

~0.5 mol dichloromethane/mol

SMILES string

[Fe].Cl[Pd]Cl.[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)N(C)C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service