Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361658

Sigma-Aldrich

Phentolamine methanesulfonate salt

99%

Synonym(s):

2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H19N3O · CH3SO3H
CAS Number:
Molecular Weight:
377.46
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

99%

mp

180-182 °C (lit.)

SMILES string

CS(O)(=O)=O.Cc1ccc(cc1)N(CC2=NCCN2)c3cccc(O)c3

InChI

1S/C17H19N3O.CH4O3S/c1-13-5-7-14(8-6-13)20(12-17-18-9-10-19-17)15-3-2-4-16(21)11-15;1-5(2,3)4/h2-8,11,21H,9-10,12H2,1H3,(H,18,19);1H3,(H,2,3,4)

InChI key

OGIYDFVHFQEFKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Useful α-adrenoceptor antagonist.

Biochem/physiol Actions

α-Adrenergic receptor antagonist; peripheral vasodilator.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Mohuczy-Dominiak et al.
The Journal of pharmacology and experimental therapeutics, 266(1), 279-287 (1993-07-01)
Marked differences in the characteristics of alpha-2 adrenoceptors in the kidney of various species have been reported. In addition, there are functional differences in alpha-2 adrenoceptors in various nephron segments of the same species. In this study we have characterized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service