Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361054

Sigma-Aldrich

2-Ethyl-2-hexenoic acid, predominantly trans

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=C(C2H5)CO2H
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100

assay

95%

refractive index

n20/D 1.46 (lit.)

bp

123-126 °C/12 mmHg (lit.)

mp

−2 °C (lit.)

density

0.95 g/mL at 25 °C (lit.)

SMILES string

CCC\C=C\C(=O)OCC

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kröger et al.
International archives of occupational and environmental health, 62(3), 213-216 (1990-01-01)
Exposure to a new wood preservative agent (Sinesto B), whose active ingredient is 2-ethylhexanoic acid (2-EHA), was determined by urinalysis of the parent chemical and its metabolites in workers employed in four Finnish sawmills. The excretion of these chemicals was
M Dunsing et al.
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 35(2), 93-95 (1989-02-01)
The carry-over of 2-ethylhexenic acid in muscle, liver and kidneys of pigs was investigated in consequence of feeding Ethexen-conserved moist grain. Residues of 2-ethylhexenic acid above the detection limit of 0.5 mg.kg-1 couldn't be established in organs and tissues by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service