Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360767

Sigma-Aldrich

2-(Methacryloyloxy)ethyl acetoacetate

95%

Synonym(s):

(2-Acetoacetoxy)ethyl methacrylate, Ethylene glycol monoacetoacetate monomethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CO2CH2CH2O2CC(CH3)=CH2
CAS Number:
Molecular Weight:
214.22
Beilstein/REAXYS Number:
2099809
EC Number:
MDL number:
UNSPSC Code:
12162002

assay

95%

contains

3,000 ppm BHT as inhibitor

refractive index

n20/D 1.456 (lit.)

bp

100 °C/0.8 mmHg (lit.)

density

1.122 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OCCOC(=O)C(C)=C

InChI

1S/C10H14O5/c1-7(2)10(13)15-5-4-14-9(12)6-8(3)11/h1,4-6H2,2-3H3

InChI key

IBDVWXAVKPRHCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Comonomer in coatings and adhesives. Binds to proteins.

Features and Benefits

Acrylic monomer. Forms chelates with metal salts. Functional groups available for condensation reactions and cross-linking. Promotes adhesion to metal surfaces.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service