Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360325

Sigma-Aldrich

Pepstatin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
Beilstein/REAXYS Number:
2201362
EC Number:
MDL number:
UNSPSC Code:
12352103

optical activity

[α]20/D −90°, c = 0.3 in methanol

mp

233 °C (dec.) (lit.)

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)[C@@H](O)CC(O)=O

InChI

1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23-,24-,25-,26-,30-,31-/m0/s1

InChI key

FAXGPCHRFPCXOO-LXTPJMTPSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Pepstatin A is an inhibitor of acid proteases (aspartyl peptidases). It forms a 1:1 complex with proteases such as pepsin, renin, cathepsin D, bovine chymosin, and protease B (Aspergillus niger). The inhibitor is highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases. Solubilized γ-secretase and retroviral protease are also inhibited by Pepstatin A. It has been used to characterize proteases from several sources.
Potent inhibitor of acid proteases, including pepsin, renin and cathepsin D and many microbial aspartic proteases. Effective concentration 1 μM. Stable for about 1 day.

Preparation Note

Pepstatin can be dissolved at 1 mg/ml in 10% (v/v) acetic acid in methanol (9:1 methanol:acetic acid). The inclusion of acetic acid may be necessary to dissolve this peptide in methanol or DMSO. It has been dissolved at 1 to 2 mg/ml in ethanol, but heat may be required for complete dissolution. Solutions of Pepstatin A can be heated as high as 60 °C without any decomposition of the peptide.
Stock solutions at 1 mg/ml are stable at least a week at 4 °C. A 1 mM solution in methanol or DMSO is stable for months at -20 °C. If solutions become darker yellow, the reagent is hydrolyzing.
A working concentration of 1 μM is stable for at least one day at room temperature.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service