Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360236

Sigma-Aldrich

1-Methyluric acid

99%

Synonym(s):

1-Methyl-2,6,8-trihydroxypurine, 1-U, 1MUA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O3
CAS Number:
Molecular Weight:
182.14
Beilstein/REAXYS Number:
195190
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

99%

SMILES string

CN1C(=O)NC2=C(NC(=O)N2)C1=O

InChI

1S/C6H6N4O3/c1-10-4(11)2-3(9-6(10)13)8-5(12)7-2/h1H3,(H,9,13)(H2,7,8,12)

InChI key

QFDRTQONISXGJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boris Rocić et al.
Bioorganic & medicinal chemistry letters, 15(4), 1181-1184 (2005-02-03)
Uric acid (1a) suppresses basal insulin release in isolated rat pancreatic islets and inhibition of glucose-stimulated insulin secretion (GSIS) occurs right at hyperuricaemic levels (0.4 mM). Conversely, 1 mM guanidinium urate (2a) was completely ineffective, strongly suggesting that binding to
Alexander C Roy et al.
Bioorganic & medicinal chemistry letters, 20(1), 141-144 (2009-12-17)
CTP synthase (CTPS) catalyzes the conversion of UTP to CTP and is a recognized target for the development of anticancer, antiviral, and antiprotozoal agents. Xanthine and related compounds inhibit CTPS activity (IC(50)=0.16-0.58mM). The presence of an 8-oxo function (i.e., uric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service