Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359831

Sigma-Aldrich

Di-tert-amyl dicarbonate

97%

Synonym(s):

(Amoc)2O, Di-tert-amyl pyrocarbonate, Di-tert-pentyl dicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH2C(CH3)2OCO]2O
CAS Number:
Molecular Weight:
246.30
Beilstein/REAXYS Number:
2265846
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.422 (lit.)

bp

70 °C/0.05 mmHg (lit.)

density

1.006 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCC(C)(C)OC(=O)OC(=O)OC(C)(C)CC

InChI

1S/C12H22O5/c1-7-11(3,4)16-9(13)15-10(14)17-12(5,6)8-2/h7-8H2,1-6H3

InChI key

PGWKKYVDHYLHNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids not in Storage Class 3

wgk_germany

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service