Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359637

Sigma-Aldrich

3-Amino-2,3-dihydrobenzoic acid hydrochloride

97%

Synonym(s):

5-Amino-1,3-cyclohexadiene-1-carboxylic acid hydrochloride, Gabaculine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO2 · HCl
CAS Number:
Molecular Weight:
175.61
Beilstein/REAXYS Number:
4552667
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

mp

203 °C (dec.) (lit.)

SMILES string

Cl[H].NC1CC(=CC=C1)C(O)=O

InChI

1S/C7H9NO2.ClH/c8-6-3-1-2-5(4-6)7(9)10;/h1-3,6H,4,8H2,(H,9,10);1H

InChI key

OBZFLUDUSNCZKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inés Hernández-Fisac et al.
The Biochemical journal, 400(1), 81-89 (2006-07-06)
OMP (oxo-4-methylpentanoic acid) stimulates by itself a biphasic secretion of insulin whereas L-leucine requires the presence of L-glutamine. L-Glutamine is predominantly converted into GABA (gamma-aminobutyric acid) in rat islets and L-leucine seems to promote its metabolism in the 'GABA shunt'
Jehoon Yang et al.
Journal of neuroscience methods, 182(2), 236-243 (2009-06-23)
Previous in vivo magnetic resonance spectroscopy (MRS) studies of gamma-aminobutyric acid (GABA) synthesis have relied on (13)C label incorporation into GABA C2 from [1-(13)C] or [1,6-(13)C(2)]glucose. In this study, the [(13)C]GABA C1 signal at 182.3 ppm in the carboxylic/amide spectral
Masahiro Irifune et al.
Canadian journal of anaesthesia = Journal canadien d'anesthesie, 54(12), 998-1005 (2007-12-07)
gamma-Aminobutyric acid (GABA) and N-methyl-D-aspartate (NMDA) receptors are important targets for anesthetic action at the in vitro cellular level. Gabaculine is a GABA-trans-aminase inhibitor that increases endogenous GABA in the brain, and enhances GABA activity. We have recently shown that
P J Loida et al.
Archives of biochemistry and biophysics, 372(2), 230-237 (1999-12-22)
The metabolite 5-aminolevulinic acid (ALA) is an early committed intermediate in the biosynthetic pathway of heme and chlorophyll formation. In plants, 5-aminolevulinic acid is synthesized via a two-step pathway in which glutamyl-tRNA(Glu) is reduced by glutamyl-tRNA(Glu) reductase (GluTR) to glutamate
Christopher Sayer et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 7), 763-772 (2012-07-04)
The three-dimensional structure of the Sulfolobus solfataricus serine:pyruvate aminotransferase has been determined to 1.8 Å resolution. The structure of the protein is a homodimer that adopts the type I fold of pyridoxal 5'-phosphate (PLP)-dependent aminotransferases. The structure revealed the PLP cofactor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service