Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359580

Sigma-Aldrich

(8S,9R)-(−)-N-Benzylcinchonidinium chloride

98%

Synonym(s):

N-Benzylcinchonidinium chloride, BCDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29ClN2O
CAS Number:
Molecular Weight:
420.97
Beilstein/REAXYS Number:
5421832
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

powder

optical activity

[α]20/D −180°, c = 1.3 in H2O

mp

210 °C (dec.) (lit.)

functional group

hydroxyl
phenyl

SMILES string

[Cl-].O[C@@H]([C@@H]1C[C@@H]2CC[N+]1(C[C@@H]2C=C)Cc3ccccc3)c4ccnc5ccccc45

InChI

1S/C26H29N2O.ClH/c1-2-20-18-28(17-19-8-4-3-5-9-19)15-13-21(20)16-25(28)26(29)23-12-14-27-24-11-7-6-10-22(23)24;/h2-12,14,20-21,25-26,29H,1,13,15-18H2;1H/q+1;/p-1/t20-,21-,25-,26+,28?;/m0./s1

InChI key

FCHYSBWCOKEPNQ-IOPLZPHGSA-M

Other Notes

Chiral phase transfer catalyst[1][2];resolutions by crystallization of the inclusion complex[3][4]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Toda et al.
The Journal of Organic Chemistry, 59, 5748-5748 (1994)
C.M. Gasparski, M.J. Miller
Tetrahedron, 47, 5367-5367 (1991)
J.H. Zhang et al.
Tetrahedron Asymmetry, 13, 1363-1363 (2002)
S. Julia et al.
Journal of the Chemical Society. Perkin Transactions 1, 574-574 (1981)

Articles

Asymmetric phase-transfer catalysis (PTC) has been recognized as a “green” alternative to many homogeneous synthetic organic transformations, and has found widespread application. Synthetically modified cinchona alkaloids are typical chiral organocatalysts used in asymmetric PTC.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service