Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

358843

Sigma-Aldrich

1,2-Dichloroethyl ethyl ether

technical grade, 85%

Synonym(s):

1,2-Dichloro-1-ethoxyethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CHClOCH2CH3
CAS Number:
Molecular Weight:
143.01
Beilstein/REAXYS Number:
1733607
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical grade

assay

85%

refractive index

n20/D 1.442 (lit.)

bp

140-145 °C (lit.)

density

1.167 g/mL at 25 °C (lit.)

SMILES string

CCOC(Cl)CCl

InChI

1S/C4H8Cl2O/c1-2-7-4(6)3-5/h4H,2-3H2,1H3

InChI key

NNBUKAPOVBEMNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service