358797
2,4,6-Tris(pentadecafluoroheptyl)-1,3,5-triazine
98%
Synonym(s):
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine
About This Item
Recommended Products
assay
98%
mp
26-29 °C (lit.)
storage temp.
2-8°C
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)c1nc(nc(n1)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C24F45N3/c25-4(26,7(31,32)10(37,38)13(43,44)16(49,50)19(55,56)22(61,62)63)1-70-2(5(27,28)8(33,34)11(39,40)14(45,46)17(51,52)20(57,58)23(64,65)66)72-3(71-1)6(29,30)9(35,36)12(41,42)15(47,48)18(53,54)21(59,60)24(67,68)69
InChI key
BFAUAMWYFFXWLX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1
target_organs
Liver
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service