Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

357219

Sigma-Aldrich

Molybdenum

foil, thickness 0.5 mm, ≥99.9% trace metals basis

Synonym(s):

Molybdenum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mo
CAS Number:
Molecular Weight:
95.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

≥99.9% trace metals basis

form

foil

resistivity

5.0 μΩ-cm, 20°C

thickness

0.5 mm

bp

4612 °C (lit.)

mp

2617 °C (lit.)

density

10.3 g/mL at 25 °C (lit.)

SMILES string

[Mo]

InChI

1S/Mo

InChI key

ZOKXTWBITQBERF-UHFFFAOYSA-N

Quantity

12.8 g = 50 × 50 mm; 51.2 g = 100 × 100 mm, 115.2 g = 150 × 150 mm

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear nonproliferation. Iranian 'medical' reactor stokes search for alternatives.
Richard Stone
Science (New York, N.Y.), 340(6139), 1392-1392 (2013-06-22)
Elena Vigonsky et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(14), 5440-5445 (2013-03-21)
In all kingdoms of life, ATP Binding Cassette (ABC) transporters participate in many physiological and pathological processes. Despite the diversity of their functions, they have been considered to operate by a largely conserved mechanism. One deviant is the vitamin B12
Phillip Ringel et al.
The Journal of biological chemistry, 288(20), 14657-14671 (2013-03-30)
Nitrate reductase (NR) is a complex molybdenum cofactor (Moco)-dependent homodimeric metalloenzyme that is vitally important for autotrophic organism as it catalyzes the first and rate-limiting step of nitrate assimilation. Beside Moco, eukaryotic NR also binds FAD and heme as additional
Joyee Mitra et al.
Inorganic chemistry, 52(6), 3032-3042 (2013-03-07)
A series of [Mo(IV)O(mnt)(SR)(N-N)](-) (mnt = maleonitriledithiolate; R = Ph, nap, p-Cl-Ph, p-CO2H-Ph, and p-NO2-Ph; N-N = 2,2'-bipyridine (bipy) and 1,10-phenanthroline (phen)) complexes analogous to the reduced active site of enzymes of the sulfite oxidase family has been synthesized and
Alexander O Terent'ev et al.
Organic & biomolecular chemistry, 11(16), 2613-2623 (2013-03-01)
Phosphomolybdic acid (PMA) and phosphotungstic acid (PTA) efficiently catalyze the addition of H2O2 to β-diketones to form bridged 1,2,4,5-tetraoxanes. These reactions are not accompanied by the formation of monocyclic peroxides containing hydroxy and hydroperoxide groups or polymeric peroxides. The use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service