Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35411

Supelco

Tetramethylammonium hydroxide solution

0.95-1.05 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(OH)
CAS Number:
Molecular Weight:
91.15
Beilstein/REAXYS Number:
3558708
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

concentration

0.95-1.05 M
~1 M in H2O

SMILES string

[OH-].C[N+](C)(C)C

InChI

1S/C4H12N.H2O/c1-5(2,3)4;/h1-4H3;1H2/q+1;/p-1

InChI key

WGTYBPLFGIVFAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Lin et al.
Biochemistry, 47(40), 10809-10815 (2008-09-13)
Potassium is an activator of the reaction catalyzed by homoisocitrate (HIc) dehydrogenase (HIcDH) from Saccharomyces cerevisiae with either the natural substrate, homoisocitrate, or the slow substrate isocitrate. On the basis of initial velocity studies, the selectivity of the activator site
Mei Fang et al.
Nanotechnology, 23(14), 145601-145601 (2012-03-22)
Magnetite nanoparticles have been prepared by co-precipitation using a custom-designed jet mixer to achieve rapid mixing (RM) of reactants in a timescale of milliseconds. The quick and stable nucleation obtained allows control of the particle size and size distribution via
A strategy for changing plasma pralidoxime kinetics and, perhaps, effect in organophosphorus insecticide poisoning.
Michael Eddleston et al.
Critical care medicine, 39(4), 908-909 (2011-05-27)
Aleksandr Saghyan et al.
Journal of neuroscience methods, 205(1), 110-118 (2012-01-11)
Numerous brain structures are composed of distinct layers and such stratification has a profound effect on extracellular diffusion transport in these structures. We have derived a more general form of diffusion equation incorporating inhomogeneities in both the extracellular volume fraction
Jalila Amorri et al.
Environmental science and pollution research international, 18(9), 1606-1616 (2011-05-25)
Complex organic compounds found in oil and sediments linked with a particular source (such as algae, bacteria or vascular plants) are defined as biomarkers and are useful dating indicators in organic geochemistry. This paper presents the composition of the organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service