Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34905

Sigma-Aldrich

Dibutylzinc solution

technical, ~1 M in heptane

Synonym(s):

Zincdibutyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2Zn
CAS Number:
Molecular Weight:
179.62
Beilstein/REAXYS Number:
3535191
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical

concentration

~1 M in heptane

SMILES string

CCCC[Zn]CCCC

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service