Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

348651

Sigma-Aldrich

Palladium

foil, thickness 0.05 mm, 99.98%

Synonym(s):

Palladium (powder), Palladium black, Palladium element

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

99.98%

form

foil

resistivity

9.96 μΩ-cm, 20°C

thickness

0.05 mm

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuxing Chen et al.
Organic & biomolecular chemistry, 11(16), 2582-2585 (2013-03-19)
A one-pot two-step synthesis of versatile indenones has been developed. This palladium(II)-catalyzed transformation involves generation and condensation of ortho-functionalized 1,2-benzils from 2-(2-arylethynylphenyl)acetonitriles using Ph2SO as the oxidant. The resulting 3-cyanoindenones can be converted to various valuable molecules.
Meifang Zheng et al.
Organic letters, 15(8), 1838-1841 (2013-04-02)
Palladium-catalyzed oxidative difunctionalization of enol ethers with 1,3-dicarbonyl compounds to construct trisubstituted furans in one step under mild conditions is described. The reaction is thought to proceed through a C-C bond formation along with a C-O bond closing the ring
Meng Wang et al.
Organic & biomolecular chemistry, 11(16), 2574-2577 (2013-03-15)
A novel Pd/Cu catalyzed domino reaction for the synthesis of functionalized pyrrolo[1,2-b]pyridazines from readily accessible (hetero)aryl propargyl alcohols and 1-amino-2-bromopyrroles was developed. This cascade process involves a Sonogashira cross-coupling reaction, an isomerization and an intramolecular condensation.
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work
V Volarevic et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 18(1), 131-137 (2013-04-25)
As novel therapeutic agents relevant to colon cancer therapy are explored continuously, we tested 4 R2edda-type ligand precursors O,O'-dialkyl esters of (S,S)-ethylenediamine-N,N'-di-2-(4-methyl)pentanoic acid (L1.2HCl-L4.2HCl) and corresponding palladium(II) and platinum(II) complexes against the human colon cancer cell lines CaCo-2, SW480 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service