346993
Palladium
rod, diam. 3.1 mm, ≥99.9% trace metals basis
Synonym(s):
Palladium (powder), Palladium black, Palladium element
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
EC Number:
MDL number:
UNSPSC Code:
12141733
PubChem Substance ID:
assay
≥99.9% trace metals basis
form
rod
resistivity
9.96 μΩ-cm, 20°C
diam.
3.1 mm
bp
2970 °C (lit.)
mp
1554 °C (lit.)
density
12.02 g/cm3 (lit.)
SMILES string
[Pd]
InChI
1S/Pd
InChI key
KDLHZDBZIXYQEI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Quantity
1.9 g = 2 cm; 9.5 g = 10 cm
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xuxing Chen et al.
Organic & biomolecular chemistry, 11(16), 2582-2585 (2013-03-19)
A one-pot two-step synthesis of versatile indenones has been developed. This palladium(II)-catalyzed transformation involves generation and condensation of ortho-functionalized 1,2-benzils from 2-(2-arylethynylphenyl)acetonitriles using Ph2SO as the oxidant. The resulting 3-cyanoindenones can be converted to various valuable molecules.
Meifang Zheng et al.
Organic letters, 15(8), 1838-1841 (2013-04-02)
Palladium-catalyzed oxidative difunctionalization of enol ethers with 1,3-dicarbonyl compounds to construct trisubstituted furans in one step under mild conditions is described. The reaction is thought to proceed through a C-C bond formation along with a C-O bond closing the ring
Meng Wang et al.
Organic & biomolecular chemistry, 11(16), 2574-2577 (2013-03-15)
A novel Pd/Cu catalyzed domino reaction for the synthesis of functionalized pyrrolo[1,2-b]pyridazines from readily accessible (hetero)aryl propargyl alcohols and 1-amino-2-bromopyrroles was developed. This cascade process involves a Sonogashira cross-coupling reaction, an isomerization and an intramolecular condensation.
Kathrin Ulbrich et al.
The Journal of organic chemistry, 78(8), 4202-4206 (2013-04-03)
Racemic 4-hydroxycyclopentenone, readily derived from furfuryl alcohol, can be transformed via its O-Boc derivative to 4-acyloxy, 4-aryloxy-, 4-amino-, or 4-thio-substituted cyclopentenones with high enantioselectivity by palladium-catalyzed kinetic resolution via nucleophilic allylic substitutions. Applying this methodology, a short formal synthesis of
Ling Li et al.
Nature chemistry, 5(7), 607-612 (2013-06-22)
The development of transition metal-catalysed cross-coupling reactions has greatly influenced the manner in which the synthesis of complex organic molecules is approached. A wide variety of methods are now available for the formation of C(sp(2))-C(sp(2)) bonds, and more recent work
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service