Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

346748

Sigma-Aldrich

Davy Reagent methyl

technical grade, 90%

Synonym(s):

2,4-Bis(methylthio)-1,3,2,4-dithiadiphosphetane-2,4-disulfide, DR-Me

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6P2S6
CAS Number:
Molecular Weight:
284.41
Beilstein/REAXYS Number:
4182442
MDL number:

grade

technical grade

assay

90%

SMILES string

CSP1(=S)SP(=S)(SC)S1

InChI

1S/C2H6P2S6/c1-7-3(5)9-4(6,8-2)10-3/h1-2H3

InChI key

OIEQWZXDRGOGHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service