Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

346527

Sigma-Aldrich

Trimethylacetic acid, anhydride with diethylborinic acid

Synonym(s):

Diethylboryl pivalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCO2B(C2H5)2
CAS Number:
Molecular Weight:
170.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

bp

60 °C/10 mmHg (lit.)

density

0.863 g/mL at 25 °C (lit.)

SMILES string

CCB(CC)OC(=O)C(C)(C)C

InChI

1S/C9H19BO2/c1-6-10(7-2)12-8(11)9(3,4)5/h6-7H2,1-5H3

InChI key

MHSAIHVCWXTSBN-UHFFFAOYSA-N

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service