Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344230

Sigma-Aldrich

Lithium 9-BBN hydride solution

1.0 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16BLi
CAS Number:
Molecular Weight:
129.96
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.919 g/mL at 25 °C

SMILES string

[Li+].[BH2-]1[C@H]2CCC[C@@H]1CCC2

InChI

1S/C8H16B.Li/c1-3-7-5-2-6-8(4-1)9-7;/h7-8H,1-6,9H2;/q-1;+1/t7-,8+;

InChI key

FKBNTWWZQZPVGU-KVZVIFLMSA-N

Application

Reactant for:
  • Preparation of Ti dihydridodiorganylborate compounds
  • Lithium imidazolium borate sulfonate triple ionic compounds as Li cation conductive matrixes
  • Reduction reactions (reducing agent)

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 2

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service