Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

343897

Sigma-Aldrich

N-Octanoyl-N-methylglucamine

98%

Synonym(s):

N-(D-Glucityl)-N-methyloctanamide, N-Methyl-N-octanoyl-D-glucamine, MEGA-8, OMEGA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H31NO6
CAS Number:
Molecular Weight:
321.41
Beilstein/REAXYS Number:
5816747
MDL number:
Pricing and availability is not currently available.

description

non-ionic

assay

98%

CMC

58 mM (20-25°C)

SMILES string

CCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C15H31NO6/c1-3-4-5-6-7-8-13(20)16(2)9-11(18)14(21)15(22)12(19)10-17/h11-12,14-15,17-19,21-22H,3-10H2,1-2H3/t11-,12+,14+,15+/m0/s1

InChI key

SBWGZAXBCCNRTM-CTHBEMJXSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service