Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

343315

Sigma-Aldrich

1-Bromo-2-ethylbenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4Br
CAS Number:
Molecular Weight:
185.06
Beilstein/REAXYS Number:
1929735
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

liquid

refractive index

n20/D 1.549 (lit.)

bp

199 °C (lit.)

density

1.338 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CCc1ccccc1Br

InChI

1S/C8H9Br/c1-2-7-5-3-4-6-8(7)9/h3-6H,2H2,1H3

InChI key

HVRUGFJYCAFAAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-2-ethylbenzene is an organobromine compound.

application

1-Bromo-2-ethylbenzene may be used in the synthesis of:
  • 1-(2′-ethylphenyl)ethanol[1]
  • N-benzyl-P-(2-ethylphenyl)-P-phenylphosphinic amide[2]
  • 4-(isopropyldimethylsilyl)-2-(2-ethylphenyl)pyridine[3]
  • 5-amino-2′-ethyl -biphenyl-2-ol, used for modification of carbon paste electrode[4]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How Space-Filling Is a Pyridine Lone Pair?
Mazzanti A, et al.
European Journal of Organic Chemistry, 33, 6725-6731 (2011)
Henok H Kinfe et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 12), o3392-o3392 (2011-12-27)
In the crystal structure of the title compound, C(21)H(22)NOP, the amine H atom is involved in N-H⋯O hydrogen-bonding inter-actions, resulting in chains along the c axis. The crystal lattice is consolidated by weak inter-molecular C-H⋯π inter-actions.
Voltammetric sensor for simultaneous determination of ascorbic acid, acetaminophen, and tryptophan in pharmaceutical products.
Beitollahi H, et al.
Ionics, 1-9 (2014)
J P Payan et al.
Drug metabolism and disposition: the biological fate of chemicals, 27(12), 1470-1478 (1999-11-26)
The excretion and metabolism of neurotoxic 1,2-diethylbenzene (1, 2-DEB) was studied in male Sprague-Dawley rats after i.v. (1 mg/kg) or oral (1 or 100 mg/kg) administration of 1,2-diethyl[U-(14)C]benzene ([(14)C]1,2-DEB). Whatever the treatment, radioactivity was mainly excreted in urine (65-76% of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service