Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

343293

Sigma-Aldrich

Zinc arsenide

−20 mesh, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn3As2
CAS Number:
Molecular Weight:
346.01
EC Number:
MDL number:
UNSPSC Code:
12352300

assay

99%

particle size

−20 mesh

mp

1015 °C

density

5.53 g/mL at 25 °C (lit.)

SMILES string

[Zn]=[As][Zn][As]=[Zn]

InChI

1S/2As.3Zn

InChI key

RHKSESDHCKYTHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service