Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

338664

Sigma-Aldrich

4′-(Heptyloxy)-4-biphenylcarbonitrile

liquid crystal (nematic), 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OC6H4C6H4CN
CAS Number:
Molecular Weight:
293.40
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

form

liquid crystal (nematic)

mp

54-57 °C (lit.)

transition temp

crystalline phase to nematic phase 54-57 °C
nematic phase to isotropic phase 75 °C

SMILES string

CCCCCCCOc1ccc(cc1)-c2ccc(cc2)C#N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service