Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

338281

Sigma-Aldrich

Bromomethane solution

2.0 M in tert-butyl methyl ether, anhydrous

Synonym(s):

Methyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Br
CAS Number:
Molecular Weight:
94.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

anhydrous

concentration

2.0 M in tert-butyl methyl ether

impurities

<0.005% water

density

0.86 g/mL at 25 °C

application(s)

agriculture

SMILES string

CBr

InChI

1S/CH3Br/c1-2/h1H3

InChI key

GZUXJHMPEANEGY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Please view www.aldrich.com/epaods regarding the EPA′s request for application information of Ozone Depleting Substances

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Ozone 1 - Skin Irrit. 2 - STOT RE 2

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-29.2 °F - closed cup

flash_point_c

-34 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lygia T Budnik et al.
Environmental health : a global access science source, 11, 5-5 (2012-01-31)
Although ozone-depleting methyl bromide was destined for phase-out by 2005, it is still widely applied as a consequence of various critical-use-exemptions and mandatory international regulations aiming to restrict the spread of pests and alien species (e.g. in globalized transport and
Alan V Barak et al.
Journal of economic entomology, 104(1), 69-74 (2011-03-17)
Ash (Fraxinus spp.) logs, infested with fully developed, cold-acclimated larval and prepupal emerald ash borer, Agrilus planipennis Fairmaire (Coleoptera: Buprestidae), were fumigated with methyl bromide (MeBr) at 4.4 and 10.0 degrees C for 24 h. Concentrations X time dosages of
Paul E Stromberg et al.
Clinical toxicology (Philadelphia, Pa.), 51(3), 182-185 (2013-03-12)
Methyl bromide is a halogenated aliphatic hydrocarbon that exists as a colorless gas or a volatile liquid. Methyl bromide historically had been used in fire extinguishers but is more commonly used as a gas fumigant for soil-borne diseases and pests.
Marcus Schortemeyer et al.
Journal of economic entomology, 104(3), 717-731 (2011-07-09)
Following the increasing international phasing out of methyl bromide for quarantine purposes, the development of alternative treatments for timber pests becomes imperative. The international accreditation of new quarantine treatments requires verification standards that give confidence in the effectiveness of a
Ting Bin Lim et al.
Journal of the American Chemical Society, 133(30), 11534-11539 (2011-06-24)
We describe the dissociative attachment (DA) of methyl bromide to form chemisorbed CH(3) and Br on a Si(100)-2×1 surface at 270 K. The patterns of DA were studied experimentally by ultra-high vacuum scanning tunneling microscopy (STM) and interpreted by ab

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service