Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

338087

Sigma-Aldrich

2′,5′-Dideoxycytidine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O3
CAS Number:
Molecular Weight:
211.22
MDL number:

assay

99%

SMILES string

O[C@H]1C[C@H](N2C(N=C(N)C=C2)=O)O[C@@H]1C

InChI

1S/C9H13N3O3/c1-5-6(13)4-8(15-5)12-3-2-7(10)11-9(12)14/h2-3,5-6,8,13H,4H2,1H3,(H2,10,11,14)/t5-,6+,8-/m1/s1

InChI key

LWMIPKUFUQWBMO-GKROBHDKSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service