Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

337870

Sigma-Aldrich

Thallium(I) cyclopentadienide

technical grade

Synonym(s):

5-Cyclopentadienyl)thallium, Cyclopentadienylthallium, NSC 114719, Thallous cyclopentadienide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5Tl
CAS Number:
Molecular Weight:
269.48
EC Number:
MDL number:
UNSPSC Code:
12000000

grade

technical grade

mp

300 °C (lit.)

SMILES string

[Tl].[CH]1[CH][CH][CH][CH]1

InChI

1S/C5H5.Tl/c1-2-4-5-3-1;/h1-5H;

InChI key

RKLJDPOVAWGBSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent in preparation of
  • η5-Cyclopentadienylpalladium(II) complexes as catalysts for vinyl addition polymerization and copolymerization reactions
  • Versatile carbon ligands via oxidative cyclization
  • Isomeric dinuclear dicyclopentadienyl Ti chloride complexes containing bridging cyclopentadienylsiloxo ligands with Me Li and Lewis acids
  • Bromide complexes of the triple-decker type
  • Naphthylazo cyclopalladates via regiospecific naphthyl carbon-hydrogen bond activation

Reactant fro:
  • Selective reduction of the U3+ metallocene amidinate

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service