About This Item
Recommended Products
assay
98%
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
96-101 °C (lit.)
functional group
phosphine
SMILES string
C(P(Cc1ccccc1)Cc2ccccc2)c3ccccc3
InChI
1S/C21H21P/c1-4-10-19(11-5-1)16-22(17-20-12-6-2-7-13-20)18-21-14-8-3-9-15-21/h1-15H,16-18H2
InChI key
IFXORIIYQORRMJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Preparation of coumarin derivatives via nickel-catalyzed cycloaddition
- Copolymerization reactions
- Rh-catalyzed hydroformylation reactions
- Platinum-catalyzed oxidative hydroxylation reactions
- Preparation of substituted allylamines by three-component coupling of alkynes, imines and organoboranes
- Ruthenium catalyzed metathesis reactions (Degenerate ligand exchange)
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service