Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

335398

Sigma-Aldrich

(3S-cis)-(+)-Tetrahydro-3-isopropyl-7a-phenylpyrrolo[2,1-b]oxazol-5(6H)-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO2
CAS Number:
Molecular Weight:
245.32
MDL number:
UNSPSC Code:
12352103

assay

98%

optical activity

[α]25/D +70°, c = 1.5 in chloroform

mp

56-58 °C (lit.)

SMILES string

CC(C)[C@H]1CO[C@@]2(CCC(=O)N12)c3ccccc3

Application

Chiral bicyclic lactams such as this are useful templates for the asymmetric synthesis of a variety of natural and unnatural products. High levels of asymmetric induction have been obtained in cycloadditions, alkylations, conjugate additions, annulations, and dihydroxylations.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Munchhoff, M.J. Meyers, A.I.
The Journal of Organic Chemistry, 60, 7084-7084 (1995)
Fray, A.H. Meyers, A.I.
The Journal of Organic Chemistry, 61, 3362-3362 (1996)
Sandham, D.A. Meyers, A.I.
Journal of the Chemical Society. Chemical Communications, 2511-2511 (1995)
A Rapid and Efficient Approach to Chiral, Nonracemic Aza Sugars from Nonsugars. A Formal Synthesis of 1,4-Dideoxy-1,4-imino-D-lyxitol.
A. I. Meyers et al.
The Journal of organic chemistry, 61(8), 2586-2587 (1996-04-19)
Brengel, G.P. Meyers, A.I.
The Journal of Organic Chemistry, 61, 3230-3230 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service